Human recombinant PTP1C (from E. coli)
Fournisseur: ProSci
PRSI91-148EA 789 EUR InStock
PRSI91-148
Human recombinant PTP1C (from E. coli)
Protéines et Peptides
Protein-Tyrosine Phosphatase 1C (PTP1C) belongs to the protein-tyrosine phosphatase family.which is known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. PTP1C is highly expressed in leukocyte cell type. It contains two SH2 domains and one tyrosine-protein phosphatase domain. The SH2 regions may interact with other cellular components to modulate its own phosphatase activity against interacting substrates. In addition, PTP1C also modulates signaling by tyrosine phosphorylated cell surface receptors.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité