Human recombinant retinol binding 4 (from E. coli)
Fournisseur: ProSci
PRSI91-149EA 490 EUR InStock
PRSI91-149
Human recombinant retinol binding 4 (from E. coli)
Protéines et Peptides
Retinol Binding Protein 4 (RBP4) is a member of the Lipocalin family and in the blood. RBP4 is the specific vector for retinol. RBP4 is expressed and secreted by adipose tissue, and is associated with insulin resistance. RBP4 delivers retinol from the liver stores to the peripheral tissues. In plasma, the RBP-retinol complex interacts with transthyretin to prevents its loss by filtration through the kidney glomeruli. Defects in RBP4 cause retinol-binding protein deficiency and can cause night vision problems.
  • This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité