Human recombinant Thymopoietin (from E. coli)
Fournisseur: ProSci
PRSI91-160EA 584 EUR InStock
PRSI91-160
Human recombinant Thymopoietin (from E. coli)
Protéines et Peptides
Thymopentin is a member of the LEM family. Thymopentin is expressed in many tissues, highly in the adult thymus and fetal liver. The N-terminal contains two structurally independent domains, LEM domain and LEM-like domain. The C-terminal domain forms a four-stranded coiled coil. Thymopentin may be involved in the structural organization of the nucleus and in the post-mitotic nuclear assembly. It is associated with T-cell development and function. Meantime, Thymopentin plays an important role, together with LMNA, in the nuclear anchorage of RB1. Thymopoietin is participated in the induction of CD90 in the thymus.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité