Human recombinant UGRP1 (from E. coli)
Fournisseur: ProSci
PRSI91-170EA 584 EUR InStock
PRSI91-170
Human recombinant UGRP1 (from E. coli)
Protéines et Peptides
Uteroglobin-Related Protein 1 (UGRP1) belongs to the secretoglobin family which has been suggested to play a role in lung inflammation and allergic diseases. UGRP1 is a 17 kDa secreted homodimeric protein that shows amino acid sequence similarity with uteroglobin. UGRP1 is expressed predominantly in the lung and low levels of expression are detected in the thyroid. Expression of UGRP1 in lung epithelial cells is enhanced by IL-10 and decreased through the activities of IL-9 and IL-5. UGRP1 interacts with the macrophage scavenger receptor with collagenous structure which is expressed by alveolar macrophages in the lung. It have suggested that UGRP1 may be involved in inflammation and pathogen clearance in the lung by binding to its receptor.
  • This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité