Human recombinant USP14 (from E. coli)
Fournisseur: ProSci
PRSI91-171EA 565 EUR InStock
PRSI91-171
Human recombinant USP14 (from E. coli)
Protéines et Peptides
Ubiquitin Carboxyl-Terminal Hydrolase 14 (USP14) belongs to the ubiquitin-specific processing (USP) family which is a deubiquitinating enzyme (DUB) with His and Cys domains. USP14 located in the cytoplasm is a proteasome-associated deubiquitinase which releases ubiquitin from the proteasome targeted ubiquitinated proteins. USP14 acts also as a physiological inhibitor of endoplasmic reticulum-associated degradation (ERAD) under the non-stressed condition by inhibiting the degradation of unfolded endoplasmic reticulum proteins via interaction with ERN1. In addition, USP14 is indispensable for synaptic development and function at neuromuscular junctions, required for the degradation of the chemokine receptor CXCR4 which is critical for CXCL12-induced cell chemotaxis.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité