Human recombinant Myc-associated factor X (from E. coli)
Fournisseur: ProSci
PRSI91-176EA 584 EUR InStock
PRSI91-176
Human recombinant Myc-associated factor X (from E. coli)
Protéines et Peptides
Myc-Associated Factor X (MAX) is a member of the basic helix-loop-helix leucine zipper (bHLHZ) family of transcription factors. It contains 1 basic helix-loop-helix (bHLH) domain. It is found in the brain, heart, and lung at high levels while lower levels are seen in the liver, kidney, and skeletal muscle. MAX forms a sequence-specific DNA-binding protein complex with MYC or MAD which recognizes the core sequence 5'-CAC[GA]TG-3'. The MYC-MAX complex is a transcriptional activator, whereas the MAD-MAX complex is a repressor. It may repress transcription via the recruitment of a chromatin remodeling complex containing H3 'Lys-9' histone methyltransferase activity.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Commander maintenant
COMMANDE
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité