Human recombinant Annexin A8 (from E. coli)
Fournisseur: ProSci
PRSI91-189EA 584 EUR InStock
PRSI91-189
Human recombinant Annexin A8 (from E. coli)
Protéines et Peptides
Annexin A8 (ANXA8) belongs to a family of calcium-dependent membrane and phospholipid binding proteins. Annexin A8 contains 4 annexin repeats separated by linking sequences of variable lengths. A pair of annexin repeats may form one binding site for calcium and phospholipid. ANXA8 is preferentially expressed in acute promyelocytic leukemia (APL) cells, which suggests its participation in hematopoietic cell differentiation. In addition, ANXA8 is an anticoagulant protein that acts as an indirect inhibitor of the thromboplastin-specific complex, which is involved in the blood coagulation cascade.
  • This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité