Human recombinant PIN (from E. coli)
Fournisseur: ProSci
PRSI91-198EA 761 EUR InStock
PRSI91-198
Human recombinant PIN (from E. coli)
Protéines et Peptides
Human Dynein Cytoplasmic Light Chain 1 (DYNLL1) has been identified as a protein that interacts with NOS1, leading to NOS1 inhibition. NOS1 dimer is destabilized after binding DYNLL1 a conformation necessary activity, and it regulate numerous biologic processes throughits effects on nitric oxide synthase activity. DYNLL1 is widely expressed, with higher expression in testis and moderate expression in brain.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité