Human Recombinant GGACT (from E. coli)
Fournisseur: ProSci
PRSI91-208EA 789 EUR InStock
PRSI91-208
Human Recombinant GGACT (from E. coli)
Protéines et Peptides
Gamma-Glutamylaminecyclotransferase is an enzyme that converts gamma-glutamylamines to free amines and 5-oxoproline which belongs to the gamma-glutamylcyclotransferase family. It shows high activity toward gamma-glutamyl-epsilon-lysine, derived from the breakdown of fibrin and contributes to degradation of proteins cross-linked by transglutaminases. It degrades the cross-link between a lysine and a glutamic acid residue from two proteins that have been cross-linked by transglutaminases. This protein adopts the newly identified cyclotransferase fold, observed in Gamma-Glutamylcyclotransferase, an enzyme with activity toward gamma-glutamyl-alpha-amino acids.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité