Human recombinant IL33 (from E. coli)
Fournisseur: ProSci
PRSI91-213EA 761 EUR InStock
PRSI91-213
Human recombinant IL33 (from E. coli)
Protéines et Peptides
Interleukin-33 (IL-33) belongs to the IL-1 superfamily. IL33 is highly expressed in endothelial venules found in tonsils, Peyer patches and mesenteric lymph nodes, but almost undetectable in placenta. IL33 induces the production of T helper-2 (Th2)-associated cytokines. IL-33 is a cytokine that mediates its biological effects by binding to and signals through IL1RL1/ST2 and IL-1 Receptor Accessory Protein (IL1RAP), activating intracellular molecules in the NF-κB and MAP kinase signaling pathways that drive production of type 2 cytokines from polarized Th2 cells.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Commander maintenant
COMMANDE
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité