Human recombinant prefoldin subunit 2 (from E. coli)
Fournisseur: ProSci
PRSI91-234EA 761 EUR InStock
PRSI91-234
Human recombinant prefoldin subunit 2 (from E. coli)
Protéines et Peptides
Prefoldin Subunit 2 (PFDN2) belongs to the Prefoldin Beta subunit family. The PFDN2 protein is one of six subunits of Prefoldin that act as a molecular chaperone complex that binds and stabilizes newly synthesized polypeptides allowing them to fold correctly. PFDN2 binds specifically to Cytosolic Chaperonin (c-CPN) and transfers target proteins to it. PFDN2 also binds to a nascent polypeptide chain and promotes folding in settings where there are many competing pathways for non-native proteins.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Commander maintenant
COMMANDE
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité