Human recombinant SNAP alpha (from E. coli)
Fournisseur: ProSci
PRSI91-255EA 761 EUR InStock
PRSI91-255
Human recombinant SNAP alpha (from E. coli)
Protéines et Peptides
alpha-Soluble NSF Attachment Protein (SNAP- alpha) is a member of the SNAP (Soluble NSF Attachment Protein) family. SNAP- alpha interacts with PRKCABP and disrupts the interaction between GRIA2 and PRKCABP, leading to the internalization of GRIA2. SNAP- alpha is required for vesicular transport between the endoplasmic reticulum and the Golgi apparatus. SNAP- alpha is in charge of the binding of NSF and therefore the formation of a 20S fusion particle.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité