Human recombinant Inhibin beta C Chain (from E. coli)
Fournisseur: ProSci
PRSI91-276EA 761 EUR InStock
PRSI91-276
Human recombinant Inhibin beta C Chain (from E. coli)
Protéines et Peptides
Inhibins/activins are involved in regulating a number of diverse functions such as hypothalamic and pituitary hormone secretion, gonadal hormone secretion, germ cell development and maturation, erythroid differentiation, insulin secretion, nerve cell survival, embryonic axial development or bone growth, depending on their subunit composition. Inhibins appear to oppose the functions of activins,Inhibins and activins inhibit and activate, respectively, the secretion of follitropin by the pituitary gland.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité