Human Recombinant NDRG1 (from E. coli)
Fournisseur: ProSci
PRSI91-279EA 761 EUR InStock
PRSI91-279
Human Recombinant NDRG1 (from E. coli)
Protéines et Peptides
Protein NDRG1 is a member of the N-Myc Downregulated Gene family, which is part of the alpha/ beta Hydrolase superfamily. Protein NDRG1 is a cytoplasmic protein that is involved in stress responses, hormone responses, cell growth and differentiation. Protein NDRG1 is necessary for p53-mediated caspase activation and apoptosis. Protein NDRG1 mutuations are reported to be the cause for hereditary motor and sensory neuropathy-Lom. Decreased NDRG1 expression in glioma is linked to tumor progression; overexpression of NDRG1 is connected to malignant status of esophageal cancer.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité