Human recombinant CD46 (from cells)
Fournisseur: ProSci
PRSI91-297EA 584 EUR InStock
PRSI91-297
Human recombinant CD46 (from cells)
Protéines et Peptides
CD46 is a type I membrane protein containing four Sushi domains. CD46 is expressed by all cells except erythrocytes. CD46 has cofactor activity for inactivation of complement components C3b and C4b by serum factor I, which protects the host cell from damage by complement. It may be involved in the fusion of the spermatozoa with the oocyte during fertilization. CD46 also acts as a costimulatory factor for T-cells which induces the differentiation of CD4+ into T-regulatory 1 cells. T-regulatory 1 cells suppress immune responses by secreting interleukin-10, and therefore are thought to prevent autoimmunity. A number of viral and bacterial pathogens exploit this property and directly induce an immunosuppressive phenotype in T-cells by binding to CD46. CD46 acts as a receptor for the Edmonston strain of measles virus, human herpesvirus-6, and type IV pili of pathogenic Neisseria.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité