Human recombinant Endocan (from cells)
Fournisseur: ProSci
PRSI91-313EA 490 EUR InStock
PRSI91-313
Human recombinant Endocan (from cells)
Protéines et Peptides
ESM-1, short from Endothelial cell-specific molecule 1, is expressed in lung, on the vascular capillary network within alveolar walls, and also at lower level in kidney. It is a secretory protein, and can be inducted by TNF and IL1B/interleukin-1beta, but not IL4/interleukin-4. This protein plays great roles in angiogenesis, sprouting, and may have potent implications in lung endothelial cell-leukocyte interactions.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité