Human recombinant Endothelial cell adhesion molecule (from cells)
Fournisseur: ProSci
PRSI91-316EA 471 EUR InStock
PRSI91-316
Human recombinant Endothelial cell adhesion molecule (from cells)
Protéines et Peptides
Endothelial Cell Adhesion Molecule (ESAM) is a 55 kDa type I transmembrane glycoprotein member of the JAM family of immunoglobulin superfamily molecules. The 390 amino acid Human ESAM contains a 216 amino acid extracellular domain (ECD) with a V-type and a C2-type immunoglobulin (Ig) domain. ESAM is specifically expressed at endothelial tight junctions and on activated platelets and performs homophilic adhesion activity. The adaptor protein membrane-associated guanylate kinase MAGI-1 has been identified as an intracellular binding partner of ESAM. In addition, ESAM at endothelial tight junctions participates in the migration of neutrophils through the vessel wall, possibly by influencing endothelial cell contacts. ESAM-deficient mice were described with lowered angiogenic potential, and accordingly, overexpression of ESAM is closely associated with certain tumor growth and metastasis. ESAM is expressed on endothelial cells, activated platelets and megakaryocytes. The ECD of human and mouse ESAM share 69% amino acid identity.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Commander maintenant
COMMANDE
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité