Human recombinant protein FAM3C (from cells)
Fournisseur: ProSci
PRSI91-317EA 584 EUR InStock
PRSI91-317
Human recombinant protein FAM3C (from cells)
Protéines et Peptides
FAM3C, also called interleukin-like EMT inducer, usually exist in most secretory epithelia. It belongs to the FAM3 family according to their sequence similarities. The up-regulation and/or mislocalization in breast cancer and liver carcinoma cells of FAM3C is strongly correlated with metastasis formation and survival. FAM3C can be involved in retinal laminar formation and promote epithelial to mesenchymal transition.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité