Human recombinant FLRT2 (from cells)
Fournisseur: ProSci
PRSI91-322EA 490 EUR InStock
PRSI91-322
Human recombinant FLRT2 (from cells)
Protéines et Peptides
Fibronectin Leucine Rich Transmembrane protein 2 (FLRT2) is a member of the fibronectin leucine rich transmembrane protein (FLRT) family. The three fibronectin leucine-rich repeat transmembrane (FLRT) proteins: FLRT1, FLRT2 and FLRT3, all contain 10 leucine-rich repeats (LRR), a type III fibronectin (FN) domain, followed by the transmembrane region, and a short cytoplasmic tail. FLRT proteins have dual properties as regulators of cell adhesion and potentiators of fibroblast growth factor (FGF) mediated signalling. The fibronectin domain of all three FLRTs can bind FGF receptors. This binding is thought to regulate FGF signaling during development. The LRR domains are responsible for both the localization of FLRTs in areas of cell contact and homotypic cell cell association. FLRT2 is expressed in a subset of the sclerotome, adjacent to the region that forms the syndetome, suggesting its involvement in the FGF signalling pathway.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité