Human recombinant CLEC4E (from cells)
Fournisseur: ProSci
PRSI91-525EA 584 EUR InStock
PRSI91-525
Human recombinant CLEC4E (from cells)
Protéines et Peptides
C-Type Lectin Domain Family 4 Member E (CLEC4E) is a 219 amino acid single-pass type II membrane protein that contains one C-type Lectin domain. It is expressed in monocytes, CLEC4E functions as a downstream target of C/EBP beta and is thought to play a role in the inflammatory response, possibly via transcriptional control of C/EBP beta . CLEC4E may play a role in the response to inflammatory stimuli in peritoneal macrophages and may be involved in immune surveillance processes under transcriptional control of CEBPB. Human CLEC4E shares 67% sequence identity with its mouse counterpart, suggesting a similar function between species. CLEC-4E exists as multiple alternatively spliced isoforms that are encoded by a gene which maps to a natural killer gene complex region on human chromosome 12.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Commander maintenant
COMMANDE
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité