Human recombinant nectin-3 (from cells)
Fournisseur: ProSci
PRSI91-544EA 490 EUR InStock
PRSI91-544
Human recombinant nectin-3 (from cells)
Protéines et Peptides
Nectin-3 is a type I transmembrane glycoprotein that belongs to the nectin family. Its precursor is 549 amino acids in length and contains an extended signal sequence of 57 amino acids, an extracellular domain (ECD) of 347 amino acids, a transmembrane segment of 21 amino acids, and a cytoplasmic region of 124 amino acids. It is predominantly expressed in testis and placenta as well as in various cell lines, including epithelial cell lines. Nectin-3 plays a role in cell-cell adhesion through heterophilic trans-interactions with nectin-like proteins or nectins, such as trans-interaction with PVRL2/Nectin-2 at Sertoli-spermatid junctions. Nectin-3 is also involved in the formation of cell-cell junctions, including adherens junctions and synapses.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité