Human Recombinant Glucagon (from Cells)
Fournisseur: ProSci
PRSI91-560EA 789 EUR InStock
PRSI91-560
Human Recombinant Glucagon (from Cells)
Protéines et Peptides
Glucagon is a secreted protein and belongs to the glucagon family. Glucagon can be cleved into 8 chains, playing an important role in initiating and maintaining hyperglycemic conditions in diabetes. Glucagon can regulates blood glucose by decreasing glycolysis and increasing gluconeogenesis. In addition, Glucagon is involved in initiating and maintaining hyperglycemic conditions in diabetes. Glucagon release is stimulated by hypoglycemia and inhibited by hyperglycemia, insulin, and somatostatin. In the glucagon antagonist, His-53 and Phe-58 are missing. This antagonist has been successfully utilized to reduce glucose concentration in vivo.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité