Human recombinant ST6GalNAc2 (from cells)
Fournisseur: ProSci
PRSI91-562EA 789 EUR InStock
PRSI91-562
Human recombinant ST6GalNAc2 (from cells)
Protéines et Peptides
Alpha-N-Acetylgalactosaminide alpha-2,6-Sialyltransferase 2 (ST6GALNAC2) belongs to the glycosyltransferase 29 family that adds sialic acids to the non-reducing ends of glycoconjugates. At the cell surface, these modifications play roles in cell-cell and cell-substrate interactions, bacterial adhesion and protein targeting. ST6GALNAC2 is localized to the Golgi apparatus membrane. ST6GALNAC2 is highly expressed in lactating mammary glands and the adult testis; it is expressed at lower levels in the kidney. ST6GALNAC2 can catalyze 2,6-sialylation of the Tn antigen.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité