Human recombinant Secretagogin (from cells)
Fournisseur: ProSci
PRSI91-565EA 584 EUR InStock
PRSI91-565
Human recombinant Secretagogin (from cells)
Protéines et Peptides
Secretagogin (SCGN) is a secreted calcium-binding protein that is found in the cytoplasm; a small proportion is associated with secretory granules and membrane fractions. SCGN contains six EF-hand domains, related to calbindin D-28K and calretinin. SCGN is thought to be involved in KCL-stimulated calcium flux and cell proliferation SCGN can be detected in human serum after ischemic neuronal damage. SCGN may function to negatively control growth and differentiation rates and, thus, indirectly inhibit cell replication.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Commander maintenant
COMMANDE
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité