Human recombinant LRRC3B (from cells)
Fournisseur: ProSci
PRSI91-571EA 584 EUR InStock
PRSI91-571
Human recombinant LRRC3B (from cells)
Protéines et Peptides
Leucine-Rich Repeat-Containing Protein 3B (LRRC3B) belongs to the LRRC3 family. LRRC3B is single-pass membrane protein and contains three leucine-rich repeats, one LRRCT domain, and one LRRNT domain. LRR-containing proteins, of which there are greater than 2,000, participate in many important processes, including plant and animal immunity, hormone-receptor interactions, cell adhesion, signal transduction, regulation of gene expression, and apoptosis. A number of microarray expression profiling studies on human cancers have shown that LRRC3B is down-regulated in gastric, breast, colon, testis, prostate and brain cancers, suggesting LRRC3B involvement in carcinogenesis
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité