Mouse recombinant Glypican 1
Fournisseur: ProSci
PRSI91-578EA 525 EUR InStock
PRSI91-578
Mouse recombinant Glypican 1
Protéines et Peptides
Glypican-1 is a cell membrane protein and belongs to the glypican family. The protein may act as a catalyst in increasing the rate of conversion of prion protein PRPN(C) to PRNP(Sc) via associating (via the heparan sulfate side chains) with both forms of PRPN, targeting them to lipid rafts and facilitating their interaction. It is required for proper skeletal muscle differentiation by sequestering FGF2 in lipid rafts preventing its binding to receptors (FGFRs) and inhibiting the FGF-mediated signaling.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité