Human recombinant Brother of CDO (from Cells)
Fournisseur: ProSci
PRSI91-606EA 525 EUR InStock
PRSI91-606
Human recombinant Brother of CDO (from Cells)
Protéines et Peptides
Brother of CDO is a protein that in humans is encoded by the BOC gene. CDON and BOC are cell surface receptors of the immunoglobulin (Ig)/fibronectin type III repeat family involved in myogenic differentiation. CDON and BOC are coexpressed during development, form complexes with each other in a cis fashion, and are related to each other in their ectodomains, but each has a unique long cytoplasmic tail.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité