Mouse recombinant Syntenin 1 (from E. coli)
Fournisseur: ProSci
PRSI91-618EA 761 EUR InStock
PRSI91-618
Mouse recombinant Syntenin 1 (from E. coli)
Protéines et Peptides
SDCBP, also called syntenin-1, is short for Syndecan-binding protein 1. It is expressed by the gene Sdcbp. SDCBP seems to function as an adapter protein. In adherens junctions, the protein may function to couple syndecans to cytoskeletal proteins or signaling components. Meanwhile it seems to couple transcription factor SOX4 to the IL-5 receptor (IL5RA). SDCBP also play a role in vesicular trafficking, and is required for the targeting of TGFA to the cell surface in the early secretory pathway.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité