Human recombinant VAMPB (from E. coli)
Fournisseur: ProSci
PRSI91-620EA 525 EUR InStock
PRSI91-620
Human recombinant VAMPB (from E. coli)
Protéines et Peptides
vesicle-associated membrane protein-associated protein B/C (VAPB) is presents in mitochondria-associated membranes that are endoplasmic reticulum membrane regions closely apposed to the outer mitochondrial membrane. It can form homodimer, and heterodimer with VAPA. It also interacts with VAMP1, VAMP2, HCV NS5A, NS5B, ZFYVE27 and RMDN3. VAPB participates in the endoplasmic reticulum unfolded protein response (UPR) by inducing ERN1/IRE1 activity. It is involved in cellular calcium homeostasis regulation.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité