Mouse recombinant CD5L
Fournisseur: ProSci
PRSI91-623EA 525 EUR InStock
PRSI91-623
Mouse recombinant CD5L
Protéines et Peptides
CD5L, also known as CT-2,SP-alpha and CD5 antigen-like. It is expressed by the gene Cd5l. The protein is expressed in thymus, liver, spleen and lymph nodes. CD5L contains 3 SRCR domains, which are 102, 101, and 103aa.long. CD5L is glycosylated during post-translational modification. It may play a role in the regulation of the immune system, and seems to play a role as an inhibitor of apoptosis.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Commander maintenant
COMMANDE
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité