Human recombinant CDK2AP2 (from cells)
Fournisseur: ProSci
PRSI91-627EA 306 EUR InStock
PRSI91-627
Human recombinant CDK2AP2 (from cells)
Protéines et Peptides
CDK2AP2, also known as DOC1R, is short for cyclin-dependent kinase 2-associated protein 2. The gene CDK2AP2 encodes this protein that interacts with cyclin-dependent kinase 2 associated protein 1. Pseudogenes associated with this gene are located on chromosomes 7 and 9. Alternatively spliced transcript variants have been observed for this gene. It belongs to the CDK2AP family. CDK2AP1 (cyclin-dependent kinase 2-associated protein 1), corresponding to the gene doc-1 (deleted in oral cancer 1), is a tumor suppressor protein. The doc-1 gene is absent or down-regulated in hamster oral cancer cells and in many other cancer cell types. The ubiquitously expressed CDK2AP1 protein is the only known specific inhibitor of CDK2, making it an important component of cell cycle regulation during G(1)-to-S phase transition.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité