Human recombinant ACOT13 (from cells)
Fournisseur: ProSci
PRSI91-672EA 584 EUR InStock
PRSI91-672
Human recombinant ACOT13 (from cells)
Protéines et Peptides
Acyl-coenzyme A thioesterase 13, also known as Thioesterase superfamily member 2, ACOT13, THEM2 and PNAS-27, is a member of the thioesterase PaaI family. Acyl-CoA thioesterases catalyze the hydrolysis of acyl-CoAs to the free fatty acid and coenzyme A (CoASH), providing the potential to regulate intracellular levels of acyl-CoAs, free fatty acids and CoASH. THEM2 is a cytoplasmic protein and exsis in a homotetramer. THEM2 has been identified as an interacting protein of phosphatidylcholine transfer protein. THEM2 also regulates hepatic lipid and glucose metabolism.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité