Human recombinant MAG (from cells)
Fournisseur: ProSci
PRSI91-674EA 584 EUR InStock
PRSI91-674
Human recombinant MAG (from cells)
Protéines et Peptides
Human Myelin-Associated Glycoprotein,also known as MAG, Siglec-4,is a cell membrane glycoprotein that is a member of the SIGLEC family of proteins.MAG contains 4 Ig-like C2-type domains and 1 Ig-like V-type domain.MAG is believed to be involved in myelination during nerve regeneration. it is a adhesion molecule in postnatal neural development that mediates sialic-acid dependent cell-cell interactions between neuronal and myelinating cells and Preferentially binds to alpha-2,3-linked sialic acid.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité