Human recombinant progonadoliberin-2 (from cells)
Fournisseur: ProSci
PRSI91-675EA 761 EUR InStock
PRSI91-675
Human recombinant progonadoliberin-2 (from cells)
Protéines et Peptides
Progonadoliberin-2, also known as Progonadoliberin II and GNRH2, belongs to the gonadotropin-releasing hormone family. GNRH2 is specially expressed in midbrain, at significantly higher levels outside the brain (up to 30-fold). GNRH2 can be cleaved into two chains, gonadoliberin-2 and GnRH-associated peptide 2. gonadoliberin-2 regulates reproduction in females by stimulating the secretion of both luteinizing- and follicle-stimulating hormones. The proproteins produce three transcript variants, but produce the same peptide hormone.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Commander maintenant
COMMANDE
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité