Human Recombinant GALNTL1 (from Cells)
Fournisseur: ProSci
PRSI91-676EA 584 EUR InStock
PRSI91-676
Human Recombinant GALNTL1 (from Cells)
Protéines et Peptides
Putative polypeptide N-acetylgalactosaminyltransferase-like protein 1, also known as Polypeptide GalNAc transferase-like protein 1, Protein-UDP acetylgalactosaminyltransferase-like protein 1, UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase-like protein 1, GalNAC-T-like protein 1, pp-GaNTase-like protein 1 and GALNTL1, belongs to the glycosyltransferase 2 family. GALNTL1 plays an important role in the protein modification and protein glycosylation process. GALNTL1 uses the manganese and calcium ion as a cofactor, may catalyze the initial reaction in O-linked oligosaccharide biosynthesis, transfers the N-acetyl-D-galactosamine residue to a serine or threonine residue on the protein receptor.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité