Human recombinant inhibin beta C Chain (from cells)
Fournisseur: ProSci
PRSI91-677EA 761 EUR InStock
PRSI91-677
Human recombinant inhibin beta C Chain (from cells)
Protéines et Peptides
Inhibin beta C chain, also known as activin beta-C chain and INHBC, belongs to the TGF-beta family. INHBC forms a homodimeric or heterodimeric through association with alpha and beta subunits, linked by one or more disulfide bonds. Inhibins are heterodimers of one alpha and one beta subunit. Activins are homo- or heterodimers of beta subunits only. Inhibins/activins regulates many physiological processes, such as hypothalamic and pituitary hormone secretion, gonadal hormone secretion, germ cell development and maturation, erythroid differentiation, insulin secretion, nerve cell survival, embryonic axial development or bone growth and so on.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité