Human recombinant netrin G1 (from cells)
Fournisseur: ProSci
PRSI91-679EA 584 EUR InStock
PRSI91-679
Human recombinant netrin G1 (from cells)
Protéines et Peptides
Netrin-G1 (NTNG1) is a member of a conserved family of proteins that act as axon guidance cues during vertebrate nervous system development. Netrin-G1 contains one laminin EGF-like domain and one laminin N-terminal domain, Netrin-G1 is highly expressed in the thalamus, lowly in other tissue. Netrin-G1 localizes to the cell membrane. Netrin-G1 interacts with NGL1 and is glycosylated in the N-terminal. In addition, Netrin-G1 can promotesneurite outgrowth of both axons and dendrites.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité