Human recombinant serpin A4 (from cells)
Fournisseur: ProSci
PRSI91-685EA 584 EUR InStock
PRSI91-685
Human recombinant serpin A4 (from cells)
Protéines et Peptides
Serpin Peptidase Inhibitor, Clade A ( alpha-1 Antiproteinase, Antitrypsin), Member 4 (Serpin A4) is a member of the Serpin family. Serpin A4 exists as a monomer and some homodimers. Serpin A4 is expressed by the liver and secreted in plasma. Serpin A4 is a regulator of vascular homeostasis capable of controlling a wide spectrum of biological actions in the cardiovascular and renal systems. It can inhibit intracellular reactive oxygen species formation in cultured cardiac and renal cells. In addition, Serpin A4 has anti-inflammatory effect. Heparin blocks kallistatin's complex formation with tissue kallikrein and abolishes its inhibitory effect on tissue kallikrein's activity.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Commander maintenant
COMMANDE
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité