Human recombinant PACSIN2 (from cells)
Fournisseur: ProSci
PRSI91-686EA 761 EUR InStock
PRSI91-686
Human recombinant PACSIN2 (from cells)
Protéines et Peptides
Protein Kinase C and Casein Kinase Substrate in Neurons Protein 2 (PACSIN2) is a member of the PACSIN family. PACSIN2 is localized to the plasma membrane via its coiled-coil domain. PACSIN2 is widely expressed and contains one FCH domain and one SH3 domain. PACSIN2 forms homo- and hetero-aggregates with other PACSINs. PACSIN2 may play a role in vesicle formation and transport. In addition, PACSIN2 is involved in linking the actin cytoskeleton with vesicle formation by regulating tubulin polymerization.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Commander maintenant
COMMANDE
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité