Human recombinant Ameloblastin (from cells)
Fournisseur: ProSci
PRSI91-697EA 761 EUR InStock
PRSI91-697
Human recombinant Ameloblastin (from cells)
Protéines et Peptides
Ameloblastin (AMBN) is a member of the Ameloblastin family. AMBN is a secreted protein and is specially expressed in ameloblast, localizing to the Tomes processes of secretory ameloblasts and in the sheath space between rod-interrod enamel Mutations of this protein may be associated with dentinogenesis imperfect and autosomal dominant amylogenesis imperfect. Ameloblastin may play an important role in the formation and mineralization of the enamel matrix. Biochemically, it is classified as an intrinsically disordered protein (IDP). Its biological role remains largely unknown.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité