Human recombinant CD316 (from cells)
Fournisseur: ProSci
PRSI91-698EA 584 EUR InStock
PRSI91-698
Human recombinant CD316 (from cells)
Protéines et Peptides
Immunoglobulin Superfamily Member 8 (IGSF8) is a single-pass membrane protein. IGSF8 contains four Ig-like C2 type domains. The Ig-like C2-type domains 3 and 4 are required for interactions with CD81. IGSF8 may regulate proliferation and differentiation of keratinocytes. IGSF8 may participate in the regulation of neurite outgrowth and maintenance of the neural network in the adult brain. It also may play a role on integrin-dependent morphology and motility functions.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité