Human recombinant CRELD2 (from cells)
Fournisseur: ProSci
PRSI91-705EA 761 EUR InStock
PRSI91-705
Human recombinant CRELD2 (from cells)
Protéines et Peptides
Cysteine-Rich with EGF-Like Domain Protein 2 (CRELD2) is a secreted protein that is a member of the CRELD family. Human CRELD2 is synthesized as a 353 amino acid precursor protein with a signal peptide, a highly conserved domain rich in glutamic acid and tryptophan (WE) and EGF-like repeats. CRELD2 is ubiquitously expressed in many tissues. CRELD2 may interact with CHRNA4 and regulate transport of alpha4- beta 2 neuronal acetylcholine receptor. In addition, CRELD2 could be a novel mediator in regulating the onset and progression of various ER stress-associated diseases.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité