Human Recombinant High Mobility Group B2 (from Cells)
Fournisseur: ProSci
PRSI91-711EA 761 EUR InStock
PRSI91-711
Human Recombinant High Mobility Group B2 (from Cells)
Protéines et Peptides
High Mobility Group Protein B2 (HMGB2) belongs to the non-histone chromosomal high-mobility group protein family. Members of this family are chromatin-associated and widely spread in the nucleus of higher eukaryotic cells. HMGB2 contains 2 HMG box DNA-binding domains. It is associated with chromatin and has the ability to bend DNA, preferentially single-stranded DNA. It is shown that HMGB2 is able to efficiently bend DNA and form DNA circles. In addition, HMGB2 is involved in the final ligation step in DNA end-joining processes of DNA double-strand breaks repair and V(D)J recombination.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité