Human recombinant IDE (from cells)
Fournisseur: ProSci
PRSI91-717EA 789 EUR InStock
PRSI91-717
Human recombinant IDE (from cells)
Protéines et Peptides
Insulin-Degrading Enzyme (IDE) is a secreted enzyme that belongs to the peptidase M16 family. IDE is a large zinc-binding protease and cleaves multiple short polypeptides that vary considerably in sequence. IDE plays a role in the cellular breakdown of insulin, IAPP, glucagon, bradykinin, kallidin, and other peptides, and thereby plays a role in intercellular peptide signaling. IDE degrades amyloid formed by APP and IAPP. IDE may participate in the degradation and clearance of naturally secreted amyloid beta -protein by neurons and microglia. IDE, which migrates at 110 kDa during gel electrophoresis under denaturing conditions, has since been shown to have additional substrates, including the signaling peptides glucagon, TGF alpha and beta -endorphin.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité