Human recombinant Anthrax Toxin Receptor 1 (from cells)
Fournisseur: ProSci
PRSI91-743EA 584 EUR InStock
PRSI91-743
Human recombinant Anthrax Toxin Receptor 1 (from cells)
Protéines et Peptides
Anthrax Toxin Receptor 1 (ANTXR1) is a single-pass type I membrane protein that belongs to the ATR family. ANTXR1 contains one VWFA domain and binds PA through the VWA domain. ANTXR1 is highly expressed in tumor endothelial cells. ANTXR1 plays a role in cell attachment and migration. ANTXR1 interacts with extracellular matrix proteins and the actin cytoskeleton, it mediates adhesion of cells to type 1 collagen and gelatin, reorganization of the actin cytoskeleton and promotes cell spreading. It is also involved in the angiogenic response of cultured umbilical vein endothelial cells, up-regulated in cultured angiogenic umbilical vein endothelial cells. Defects in ANTXR1 are associated with susceptibility to hemangioma capillary infantile (HCI).
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité