Human Recombinant CD160 (from Cells)
Fournisseur: ProSci
PRSI91-744EA 365 EUR InStock
PRSI91-744
Human Recombinant CD160 (from Cells)
Protéines et Peptides
CD160 antigen is a Lipid-anchor that exists as a disulfide-linked homomultimer. CD160 contains one Ig-like V-type domain. The human CD160 precursor is a cysteine-rich, glycosylphosphatidylinositol-anchored protein of 181 amino acids with a single Ig-like domain. It is weakly homologous to KIR2DL4. CD160 is expressed in the spleen, peripheral blood, and small intestine. Its expression is tightly associated with peripheral blood NK cells and CD8 T lymphocytes with cytolytic effector activity. CD160 is a receptor showing broad specificity for both classical and non-classical MHC class I molecules.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Commander maintenant
COMMANDE
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité