Human recombinant IGF Binding 5 (from cells)
Fournisseur: ProSci
PRSI91-749EA 761 EUR InStock
PRSI91-749
Human recombinant IGF Binding 5 (from cells)
Protéines et Peptides
Insulin-Like Growth Factor-Binding Protein 5 (IGFBP-5) is a secreted protein that belongs to the insulin-like growth factor (IGF) binding proteins superfamily. Members of this family prolong the half-life of the IGFs. They have been shown to either inhibit or stimulate the growth promoting effects of the IGFs on cell culture. They alter the interaction of IGFs with their cell surface receptors. IGFBP-5 contains one IGFBP N-terminal domain and one thyroglobulin type-1 domain. IGFBP-5 is expressed by fibroblasts, myoblasts and Osteosarcoma. It is also present at lower levels in liver, kidney, and brain.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité