Human recombinant Calumenin (from Cells)
Fournisseur: ProSci
PRSI91-751EA 584 EUR InStock
PRSI91-751
Human recombinant Calumenin (from Cells)
Protéines et Peptides
Calumenin is a secreted calcium-binding protein that belongs to the CREC family. Calumenin contains six EF-hand domains and is expressed at high levels in the heart, placenta and skeletal muscle. Human Calumenin is synthesized as a 315 amino acid precursor that contains a 19 amino acid signal sequence, and a 296 amino acid mature chain. Calumenin localizes to the endoplasmic reticulum (ER) and sarcoplasmic reticulum (SR) of mammalian tissues which plays a role in ER functions as protein folding and sorting. Calumenin is involved in the regulation of vitamin K-dependent carboxylation of multiple N-terminal glutamate residues. It seems to inhibit gamma -carboxylase GGCX.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité