Human recombinant SECTM1 (from cells)
Fournisseur: ProSci
PRSI91-755EA 525 EUR InStock
PRSI91-755
Human recombinant SECTM1 (from cells)
Protéines et Peptides
Secreted and Transmembrane Protein 1 (SECTM1) is a transmembrane and secreted protein that belongs to the SECTM family. SECTM1 is expressed in a perinuclear Golgi-like pattern. It is detected at the highest levels in peripheral blood leukocytes and breast cancer cell lines. SECTM1 is considered to participate in thymocyte signaling and the hematopoietic/immune system processes. It is reported that SECTM1 is a broadly expressed, IFN- gamma -inducible molecule, which functions as a potent costimulatory ligand for T cell activation and is synergistic with anti-CD28.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité