Human recombinant LDL Receptor (from cells)
Fournisseur: ProSci
PRSI91-784EA 584 EUR InStock
PRSI91-784
Human recombinant LDL Receptor (from cells)
Protéines et Peptides
Low-Density Lipoprotein Receptor (LDLR) is a transmembrane glycoprotein that plays a critical role in cholesterol homeostasis. LDLR mediates blood cholesterol level by interacting with lipoprotein particles like LDL and VLDL. The extracellular domain of LDLR contains LDL receptor type A (ligand-binding) modules (LA repeats), epidermal growth factor-like modules, and LY repeats containing the YWTD consensus motif that are important in binding and releasing of ApoB-100 and ApoE in lipoprotein particles. The C terminal domain of LDLR inside the cell is required for the receptor internalization. Loss of function mutations in the LDLR gene causes Familial Hypercholesterolemia (FH).
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Commander maintenant
COMMANDE
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité